0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Bromo-2,3-difluoroaniline
4-Bromo-2,3-difluoroaniline 97%
4-Bromo-2,3-difluoroaniline 97%
:
Apollo Scientific
4-Bromo-2,3-difluoroaniline 97%
:
Apollo Scientific
:
112279-72-8
:
:
C₆H₄BrF₂N
:
MFCD09030643
:
208.0035 g/mol
:
60…62 °C
:
GHS07
:
Refrigerator