Order Entry
Export
ContactUsLinkComponent
2,4-Difluorocinnamic acid
2,4-Difluorocinnamic acid
Catalog # APOSPC49232-25G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Difluorocinnamic acid
Catalog # APOSPC49232-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :184.14 g/mol
Storage Temperature:Ambient
CAS Number:774-73-2
 :94977-52-3
Formula:C₉H₆F₂O₂
MDL Number:MFCD00010317