

Methyl trifluoroacetate 97%
Catalog # APOSPC49140-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:128.05 g/mol
Boiling Pt:43…44 °C
Melting Pt:–78 °C
Density:1.283 g/cm³ (25 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:207-074-5
UN:2924
ADR:3+8,II
:GHS02
:GHS05
CAS Number:431-47-0
Formula:C₃H₃F₃O₂
MDL Number:MFCD00000417