Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-4-nitrophenol
2,6-Difluoro-4-nitrophenol 95%
2,6-Difluoro-4-nitrophenol 95%
2,6-Difluoro-4-nitrophenol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,6-Difluoro-4-nitrophenol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
658-07-1
:
Formula:
C₆H₃F₂NO₃
MDL Number:
MFCD07366521
:
175.0900 g/mol
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III