Order Entry
Export
ContactUsLinkComponent
3,4-Difluoro-2-methylbenzonitrile 98%
Toxic
3,4-Difluoro-2-methylbenzonitrile 98%
Catalog # APOSPC48776-5G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
3,4-Difluoro-2-methylbenzonitrile 98%
Catalog # APOSPC48776-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :153.13 g/mol
Melting Pt:39…44 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:847502-83-4
Formula:C₈H₅F₂N
MDL Number:MFCD07777154