Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-4-trifluoromethyl-1H-imidazole
2-Methyl-4-trifluoromethyl-1H-imidazole 95%
2-Methyl-4-trifluoromethyl-1H-imidazole 95%
Irritant
2-Methyl-4-trifluoromethyl-1H-imidazole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Methyl-4-trifluoromethyl-1H-imidazole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
33468-67-6
:
Formula:
C₅H₅F₃N₂
MDL Number:
MFCD10000863
:
150.1 g/mol
Melting Pt:
179…180 °C
:
GHS07
Storage Temperature:
Ambient