0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,3-Bis(1,1,2,2-tetrafluoroethoxy)benzene
1,3-Bis(1,1,2,2-tetrafluoroethoxy)benzene
1,3-Bis(1,1,2,2-tetrafluoroethoxy)benzene
:
Apollo Scientific
1,3-Bis(1,1,2,2-tetrafluoroethoxy)benzene
:
Apollo Scientific
:
3914-19-0
:
:
C₁₀H₆F₈O₂
:
MFCD00497882
:
78…79 °C/10mmHg
:
GHS07
:
Ambient