Order Entry
Export
ContactUsLinkComponent
2,5-Difluorophenylhydrazine 98%
Irritant
2,5-Difluorophenylhydrazine 98%
Catalog # APOSPC4585-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,5-Difluorophenylhydrazine 98%
Catalog # APOSPC4585-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.12 g/mol
Melting Pt:73…76 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:97108-50-4
Formula:C₆H₆F₂N₂
MDL Number:MFCD00013384