
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:656-42-8
:
Formula:C₈H₄F₂O₃
MDL Number:MFCD00792420
:186.11 g/mol
Boiling Pt:202…203 °C
Density:1.422 g/cm³ (25 °C)
Flash Pt:96 °C
:GHS07
Storage Temperature:Refrigerator