Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,α,α-Trifluoro-m-xylene
α,α,α-Trifluoro-m-xylene 98%
α,α,α-Trifluoro-m-xylene 98%
:
Apollo Scientific
α,α,α-Trifluoro-m-xylene 98%
:
Apollo Scientific
:
401-79-6
:
:
C₈H₇F₃
:
MFCD00190105
:
160.14 g/mol
:
130 °C
:
1.148 g/cm³ (25 °C)
:
26 °C
:
Ambient
:
1993
:
3,III
:
GHS02
:
GHS07