Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-o-xylene
4-Fluoro-o-xylene 98%
4-Fluoro-o-xylene 98%
:
Apollo Scientific
4-Fluoro-o-xylene 98%
:
Apollo Scientific
:
452-64-2
:
:
C₈H₉F
:
MFCD00039217
:
124.16 g/mol
:
148…149 °C
:
1 g/cm³ (25 °C)
:
40 °C
:
Ambient
:
1993
:
3,III
:
GHS02
:
GHS07