
3-Fluoro-o-xylene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:443-82-3
:
Formula:C₈H₉F
MDL Number:MFCD00000323
Boiling Pt:147…152 °C
Density:0.992 g/cm³ (25 °C)
Flash Pt:36 °C
Storage Temperature:Ambient
EINECS:207-140-3
UN:1993
ADR:3,III
:124.16 g/mol
:GHS02