

2-Fluoro-m-xylene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:443-88-9
:
Formula:C₈H₉F
MDL Number:MFCD00039215
:124.16 g/mol
Boiling Pt:147…148 °C
Density:0.988 g/cm³ (25 °C)
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:207-144-5
UN:1993
ADR:3,III
:GHS02
:GHS07