Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-nitrotoluene
4,5-Difluoro-2-nitrotoluene 98%
4,5-Difluoro-2-nitrotoluene 98%
Toxic
4,5-Difluoro-2-nitrotoluene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
4,5-Difluoro-2-nitrotoluene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
127371-50-0
:
Formula:
C₇H₅F₂NO₂
MDL Number:
MFCD04973772
ADR:
6.1,III
:
173.12 g/mol
UN:
2810
:
GHS06
Storage Temperature:
Ambient