
α-Chloro-3,4-difluorotoulene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:698-80-6
:
Formula:C₇H₅ClF₂
MDL Number:MFCD00236218
:162.57 g/mol
Boiling Pt:27…29 °C/0.4mmHg
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05