2,4-Dichloro-5-fluorotoluene
Catalog # APOSPC430503-2G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:2 g
- Seq:3107
Specifications
About this item
:
:179.02 g/mol
Storage Temperature:Ambient
CAS Number:86522-86-3
Formula:C₇H₅Cl₂F
MDL Number:MFCD06246852