Order Entry
Export
ContactUsLinkComponent
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione
Catalog # APOSPC430133-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione
Catalog # APOSPC430133-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.19 g/mol
Storage Temperature:Ambient
CAS Number:329710-82-9
Formula:C₁₁H₈F₃NO₃
MDL Number:MFCD19686683