
1-Fluoropyridiniumtrifluoromethanesulphonate 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:107263-95-6
:
Formula:C₆H₅F₄NO₃S
MDL Number:MFCD00013458
:247.17 g/mol
Melting Pt:180…186 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05