Order Entry
Export
ContactUsLinkComponent
(3,3-Difluorocyclobutyl)-N-methylmethanamine hydrochloride
(3,3-Difluorocyclobutyl)-N-methylmethanamine hydrochloride
Catalog # APOSPC420015-1G
Supplier:  Apollo Scientific
CAS Number:  
(3,3-Difluorocyclobutyl)-N-methylmethanamine hydrochloride
Catalog # APOSPC420015-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.62 g/mol
Storage Temperature:Ambient
CAS Number:1250444-03-1
Formula:C₆H₁₂ClF₂N
MDL Number:MFCD16987777