Order Entry
Export
ContactUsLinkComponent
1-Amino-3,3-difluorocyclobutane-1-methanol hydrochloride
1-Amino-3,3-difluorocyclobutane-1-methanol hydrochloride
Catalog # APOSPC420010-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Amino-3,3-difluorocyclobutane-1-methanol hydrochloride
Catalog # APOSPC420010-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.59 g/mol
Storage Temperature:Ambient
CAS Number:1363380-82-8
Formula:C₅H₁₀ClF₂NO
MDL Number:MFCD23703610