
1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid
Catalog # APOSPC420009-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.23 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1363380-83-9
Formula:C₁₀H₁₅F₂NO₄
MDL Number:MFCD22415254