
Diisopropyl-3,3-difluorocyclobutane-1,1-dicarboxylate
Catalog # APOSPC420007-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.27 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1225532-89-7
Formula:C₁₂H₁₈F₂O₄
MDL Number:MFCD23105827