Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoro-6-methoxybenzaldehyde
2,3-Difluoro-6-methoxybenzaldehyde 98%
2,3-Difluoro-6-methoxybenzaldehyde 98%
Irritant
2,3-Difluoro-6-methoxybenzaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Difluoro-6-methoxybenzaldehyde 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
187543-87-9
:
Formula:
C₈H₆F₂O₂
MDL Number:
MFCD02093968
:
172.13 g/mol
Melting Pt:
57…62 °C
:
GHS07
Storage Temperature:
Ambient