
(±)-2,6-Difluoromandelic acid 90%
Catalog # APOSPC4115-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:188.13 g/mol
Melting Pt:142…145 °C
:GHS07
Storage Temperature:Ambient
CAS Number:207981-50-8
Formula:C₈H₆F₂O₃
MDL Number:MFCD00061298