Order Entry
Export
ContactUsLinkComponent
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95+%
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95+%
Catalog # APOSPC409044-250MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95+%
Catalog # APOSPC409044-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :297.3 g/mol
Storage Temperature:Ambient
CAS Number:1067915-34-7
Formula:C₁₅H₁₇F₂NO₃
MDL Number:MFCD16036533