
2-Amino-4,6-difluorobenzoic acid
Catalog # APOSPC4086-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:173.1169 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:126674-77-9
Formula:C₇H₅F₂NO₂
MDL Number:MFCD03618449