Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoro-2-methylpyrimidine-4,6-diol
5-Fluoro-2-methylpyrimidine-4,6-diol
5-Fluoro-2-methylpyrimidine-4,6-diol
APOSPC408555-1G
:
Apollo Scientific
:
5-Fluoro-2-methylpyrimidine-4,6-diol
APOSPC408555-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
144.11 g/mol
:
Ambient
:
1598-63-6
:
C₅H₅FN₂O₂
:
MFCD11520459