Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-5-fluoropyrimidin-2-amine
4,6-Dichloro-5-fluoropyrimidin-2-amine
4,6-Dichloro-5-fluoropyrimidin-2-amine
4,6-Dichloro-5-fluoropyrimidin-2-amine
Supplier:
Apollo Scientific
Certificates
Expand image
4,6-Dichloro-5-fluoropyrimidin-2-amine
Supplier:
Apollo Scientific
About this item
CAS Number:
15598-33-1
:
Formula:
C₄H₂Cl₂FN₃
MDL Number:
MFCD11870221
:
181.99 g/mol
Storage Temperature:
Ambient