Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Irritant
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Bromo-1,3-difluoro-2-(trifluoromethoxy)benzene
Supplier:
Apollo Scientific
About this item
CAS Number:
115467-07-7
:
Formula:
C₇H₂BrF₅O
MDL Number:
MFCD03412225
:
276.99 g/mol
:
GHS07
Storage Temperature:
Ambient