Order Entry
Export
ContactUsLinkComponent
4-Chloro-5,6-difluoronicotinic acid 95%
Irritant
4-Chloro-5,6-difluoronicotinic acid 95%
Catalog # APOSPC408223-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Chloro-5,6-difluoronicotinic acid 95%
Catalog # APOSPC408223-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.5400 g/mol
Melting Pt:117…118 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:851386-32-8
Formula:C₆H₂ClF₂NO₂
MDL Number:MFCD09031172