2,2-Difluoroacetamide 97%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:359-38-6
- Formula:C₂H₃F₂NO
- MDL Number:MFCD01896431
- Molecular Weight:95.05 g/mol
- Melting Pt:45…55 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:359-38-6
- Formula:C₂H₃F₂NO
- MDL Number:MFCD01896431
- Molecular Weight:95.05 g/mol
- Melting Pt:45…55 °C
- Storage Temperature:Ambient



