Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3-Difluoroazetidine hydrochloride
3,3-Difluoroazetidine hydrochloride
3,3-Difluoroazetidine hydrochloride
Irritant
3,3-Difluoroazetidine hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,3-Difluoroazetidine hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
288315-03-7
:
Formula:
C3H6ClF2N
MDL Number:
MFCD05663714
:
129.54 g/mol
:
GHS07
Storage Temperature:
Refrigerator