Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Chloro-4-(trifluoromethyl)pyridin-2-ol
6-Chloro-4-(trifluoromethyl)pyridin-2-ol
6-Chloro-4-(trifluoromethyl)pyridin-2-ol
Irritant
6-Chloro-4-(trifluoromethyl)pyridin-2-ol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Chloro-4-(trifluoromethyl)pyridin-2-ol
Supplier:
Apollo Scientific
About this item
CAS Number:
34486-07-2
:
Formula:
C₆H₃ClF₃NO
MDL Number:
MFCD13190002
:
197.55 g/mol
Melting Pt:
101…103 °C
:
GHS07
Storage Temperature:
Ambient