
1,3-difluoro-2-methyl-4-nitrobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:79562-49-5
:
Formula:C₇H₅F₂NO₂
MDL Number:MFCD11519030
:173.119 g/mol
Melting Pt:35…38 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07