Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Difluoropyridazine
3,6-Difluoropyridazine 95%
3,6-Difluoropyridazine 95%
:
Apollo Scientific
3,6-Difluoropyridazine 95%
:
Apollo Scientific
:
33097-39-1
:
:
C4H2F2N2
:
MFCD09909693
:
116.0700 g/mol
:
GHS07
:
Ambient