Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2,3-dihydro-1H-inden-1-one
4,5-Difluoro-2,3-dihydro-1H-inden-1-one 97%
4,5-Difluoro-2,3-dihydro-1H-inden-1-one 97%
APOSPC400711-1G
:
Apollo Scientific
:
4,5-Difluoro-2,3-dihydro-1H-inden-1-one 97%
APOSPC400711-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
168.1400 g/mol
:
Ambient
:
628732-11-6
:
C₉H₆F₂O
:
MFCD07772124