Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Difluoro-1-butene
1,1-Difluoro-1-butene 98%
1,1-Difluoro-1-butene 98%
:
Apollo Scientific
1,1-Difluoro-1-butene 98%
:
Apollo Scientific
:
407-09-0
:
:
C₄H₆F₂
:
MFCD04038010
:
92.09 g/mol
:
3.7 °C/760mmHg
:
–144.9 °C
:
1.0216 g/cm³ (25 °C)
:
Ambient
:
3161
:
2.1,
:
GHS02