

4-Chloro-2,5-difluorobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:286932-78-3
:
Formula:C₆H₂Cl₂F₂O₂S
MDL Number:MFCD02091386
:247.05 g/mol
UN:1760
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient