
5-Fluoro-o-toluonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77532-79-7
:
Formula:C₈H₆FN
MDL Number:MFCD00042295
:135.14 g/mol
Melting Pt:42…45 °C
Flash Pt:79 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07