Order Entry
Export
Orders LinkContactUsLinkComponent
 

Poly(vinylidene fluoride)

Irritant
Supplier:   Apollo Scientific
Poly(vinylidene fluoride)
Supplier:  Apollo Scientific
Certificates
Poly(vinylidene fluoride)

3 Options Available Below

Product Details & Documents
  • CAS Number:24937-79-9
  • Formula:(-CH₂CF₂-)n
  • MDL Number:MFCD00084470
  • Melting Pt:155…165 °C
  • Density:1.74 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,00 €
    29,00 €
    1 * 5 g
       29,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 50,70 €
    50,70 €
    1 * 25 g
       50,70 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 160,00 €
    160,00 €
    1 * 100 g
       160,00 €