Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane 70%
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane 70%
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane 70%
Supplier:
Apollo Scientific
Certificates
Expand image
1,1-Dichloro-2-iodo-1,2,2-trifluoroethane 70%
Supplier:
Apollo Scientific
About this item
CAS Number:
661-66-5
:
Formula:
C₂Cl₂F₃I
MDL Number:
MFCD02093303
:
278.83 g/mol
Density:
2.2 g/cm³ (25 °C)
Storage Temperature:
Ambient