Order Entry
Export
ContactUsLinkComponent
1,2-Dibromo-1,1-dichloro-2,2-difluoroethane 97%
Irritant
1,2-Dibromo-1,1-dichloro-2,2-difluoroethane 97%
Catalog # APOSPC3783-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,2-Dibromo-1,1-dichloro-2,2-difluoroethane 97%
Catalog # APOSPC3783-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :292.73
Boiling Pt:117 °C
Melting Pt:46 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:558-57-6
Formula:C₂Br₂Cl₂F₂
MDL Number:MFCD00039319