
2,5-Difluorobenzenesulphonamide
Catalog # APOSPC3478-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:193.17 g/mol
Melting Pt:137 °C
:GHS07
Storage Temperature:Ambient
CAS Number:120022-63-1
Formula:C₆H₅F₂NO₂S
MDL Number:MFCD00798115