Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dichloro-3-fluorobenzene
1,2-Dichloro-3-fluorobenzene 96%
1,2-Dichloro-3-fluorobenzene 96%
Irritant
1,2-Dichloro-3-fluorobenzene 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,2-Dichloro-3-fluorobenzene 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
36556-50-0
:
Formula:
C₆H₃Cl₂F
MDL Number:
MFCD00060647
:
164.9924 g/mol
Boiling Pt:
80 °C/22mmHg
:
GHS07
Storage Temperature:
Ambient