Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Divinyloctafluorobutane
1,4-Divinyloctafluorobutane
1,4-Divinyloctafluorobutane
APOSPC3118-1G
:
Apollo Scientific
:
1,4-Divinyloctafluorobutane
APOSPC3118-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
254.12 g/mol
:
61 °C/10mmHg
:
1.361 g/cm³ (25 °C)
:
Ambient
:
211-650-1
:
1993
:
3,III
:
GHS02
:
GHS07
:
678-65-9
:
C₈H₆F₈
:
MFCD00077520