
1-Bromo-2,4-bis(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:327-75-3
:
Formula:C₈H₃BrF₆
MDL Number:MFCD00074904
:293.01 g/mol
Boiling Pt:158 °C/740mmHg
Density:1.738 g/cm³ (25 °C)
Flash Pt:69 °C
:GHS07
Storage Temperature:Ambient