
3,5-Bis(trifluoromethyl)benzylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85068-29-7
:
Formula:C₉H₇F₆N
MDL Number:MFCD00009909
Boiling Pt:82…84 °C/15mmHg
Melting Pt:50…55 °C
Flash Pt:78 °C
Storage Temperature:Ambient
EINECS:285-290-9
UN:3259
ADR:8,II
:243.15 g/mol
:GHS05