
2,6-Bis(trifluoromethyl)benzoyl chloride 98%
Catalog # APOSPC3103L-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.57 g/mol
Boiling Pt:81…82 °C/10mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
CAS Number:53130-44-2
Formula:C₉H₃ClF₆O
MDL Number:MFCD00061228