
3,5-Bis(trifluoromethyl)benzophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21221-93-2
:
Formula:C₁₅H₈F₆O
MDL Number:MFCD00015510
:318.22 g/mol
Boiling Pt:111…112 °C/0.2mmHg
EINECS:244-282-5
:GHS07
Storage Temperature:Ambient