
3,3'-Bis(trifluoromethyl)benzhydrol 97%
Catalog # APOSPC3086D-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:118 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1598-89-6
Formula:C₁₅H₁₀F₆O
MDL Number:MFCD00039228